Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNMT1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA1880

Catalog No. PA1880


Only null left
Edge
Add to Cart

Description

Description

PA1-880 detects DNA methyltransferase 1 (Dmnt1) from HeLa cell nuclear extracts. PA1-880 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~180 kDa protein representing Dnmt1 from HeLa cell nuclear extract. A doublet between 40 and 45 kDa is also seen on Western blots using this antibody. The PA1-880 immunogen is a synthetic peptide corresponding to residues A(171) K G P A K R K P Q E E S E(185) of human Dnmt1. PA1-880 immunizing peptide (Cat. # PEP-136) is available for use in neutralization and control experiments.

This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DNMT1
Polyclonal
Unconjugated
DNMT1
ADCADN; AIM; CXXC9; CXXC-type zinc finger protein 9; DNA (cytosine-5-)-methyltransferase 1; DNA (cytosine-5)-methyltransferase 1; DNA methyltransferase (cytosine-5) 1; DNA methyltransferase 1; DNA methyltransferase HsaI; DNA methyltransferase I; DNA methyltransferase MmuI; DNA MTase HsaI; DNA MTase MmuI; DNA MTase RnoIP; DNMT; Dnmt1; Dnmt1o; FLJ16293; HSN1E; m.HsaI; m.MmuI; m.RnoIP; MCMT; Met1; Met-1; MGC104992; MommeD2; MTase; Uim
Rabbit
Antigen affinity chromatography
RUO
1786, 84350
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1 mg/mL BSA and 0.05% sodium azide
P26358, Q9Z330
DNMT1
Synthetic peptide corresponding to residues A(171) K G P A K R K P Q E E S E(185) of human Dnmt1.
100 μg
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.