Learn More
Invitrogen™ DNMT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120552
Description
Positive test controls include: Jurkat. The target is usually found in the following locations: Nucleus. Immunogen sequence: GDCVSVIPDD SSKPLYLARV TALWEDSSNG QMFHAHWFCA GTDTVLGATS DPLELFLVDE CEDMQLSYIH SKVKVIYKAP SENWAMEGGM DPESLLEGDD GKTYFYQLWY DQDYARFESP PKTQPTEDNK FKFCVSCARL AEMRQKEIPR VLEQLEDLDS RVLYYSATKN GILYRVGDGV YLPPEAFTFN IKLSSPVKRP RKEPVDEDLY PEHYRKYSDY IKGSNLDAPE PYRIGRIKEI FCPKKSNGRP NETDIKIRVN KFYRPENTHK STPASYHADI NLLYWSDEEA VVDFKAVQGR CTVEYGEDLP ECVQVYSMGG PNRFYFLEAY NAKSKSFEDP PNHARSPGNK.
This gene encodes a subunit of the augmin complex, which regulates centrosome and mitotic spindle integrity, and is necessary for the completion of cytokinesis. The encoded protein was identified by interaction with ubiquitin C-terminal hydrolase 37. Alternative splicing results in multiple transcript variants.
Specifications
| DNMT1 | |
| Polyclonal | |
| Unconjugated | |
| DNMT1 | |
| ADCADN; AIM; CXXC9; CXXC-type zinc finger protein 9; DNA (cytosine-5-)-methyltransferase 1; DNA (cytosine-5)-methyltransferase 1; DNA methyltransferase (cytosine-5) 1; DNA methyltransferase 1; DNA methyltransferase HsaI; DNA methyltransferase I; DNA methyltransferase MmuI; DNA MTase HsaI; DNA MTase MmuI; DNA MTase RnoIP; DNMT; Dnmt1; Dnmt1o; FLJ16293; HSN1E; m.HsaI; m.MmuI; m.RnoIP; MCMT; Met1; Met-1; MGC104992; MommeD2; MTase; Uim | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13433, 1786, 84350 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.87 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P13864, P26358, Q9Z330 | |
| DNMT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 760-1109 of DNMT1 (NP_001370.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.