Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dnmt2 Antibody (102B1259.2), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
$501.50
Specifications
Antigen | Dnmt2 |
---|---|
Clone | 102B1259.2 |
Concentration | 0.5 mg/mL |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
Dnmt2 Monoclonal specifically detects Dnmt2 in Human, Mouse, Zebrafish samples. It is validated for Western Blot, Flow Cytometry.Specifications
Dnmt2 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1 | |
TRDMT1 | |
IgG1 κ | |
Protein G purified | |
In mouse Dnmt2 transfected cell lysate, a 42 kDa band is oberseved. |
102B1259.2 | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Chromatin Modifiers, Chromatin Research, Epigenetics | |
Q6ICS7 | |
1787 | |
This antibody was raised against synthetic peptides corresponding to mouse Dnmt2. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title