Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dnmt2 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298992AF750
Description
Dnmt2 Polyclonal specifically detects Dnmt2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Dnmt2 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human Dnmt2. | |
0.1 mL | |
Chromatin Research, Epigenetics | |
1787 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction