Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNMT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557884 Supplier Invitrogen™ Supplier No. PA557884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVLELYSGVG GMHHALRESC IPAQVVAAID VNTVANEVYK YNFPHTQLLA KTIEGITLEE FDRLSFDMIL MSPPCQPFTR IGRQGDMTDS RT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.

CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a protein with similarity to DNA methyltransferases, but this protein does not display methyltransferase activity. The protein strongly binds DNA, suggesting that it may mark specific sequences in the genome. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNMT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRDMT1
Gene Accession No. O14717
Gene Alias DMNT2; DNA (cytosine-5)-methyltransferase-like protein 2; DNA cytosine-5 methyltransferase 2; DNA methyltransferase 2; DNA methyltransferase homolog HsaIIP; DNA methyltransferase homolog MmuIIP; DNA methyltransferase-2; DNA MTase homolog HsaIIP; DNA MTase homolog MmuIIP; Dnmt2; M.HsaIIP; m.MmuIIP; Met2; met-2; MHSAIIP; PUMET; RNMT1; Rnmt2; RP11-406H21.1; Trdmt1; tRNA (cytosine(38)-C(5))-methyltransferase; tRNA (cytosine-5-)-methyltransferase; tRNA aspartic acid methyltransferase 1
Gene Symbols TRDMT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNMT2. Recombinant protein control fragment (Product #RP-102756).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1787
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.