Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3A Antibody (64B814.1), Novus Biologicals™
SDP

Catalog No. p-4977157 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056521T 0.025 mg
NB10056521 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056521T Supplier Novus Biologicals Supplier No. NB10056521SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

DNMT3A Monoclonal specifically detects DNMT3A in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).

Specifications

Antigen DNMT3A
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone 64B814.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Flow (Intracellular)
Gene Accession No. Q9Y6K1
Gene Alias DNA (cytosine-5-)-methyltransferase 3 alpha, DNA (cytosine-5)-methyltransferase 3A, DNA cytosine methyltransferase 3A2, DNA methyltransferase HsaIIIA, DNA MTase HsaIIIA, Dnmt3a, DNMT3A2, EC 2.1.1.37, M.HsaIIIA
Gene Symbols DNMT3A
Host Species Mouse
Immunogen This antibody was raised against bacteria expressed HIS-tag recombinant mouse Dnmt3a.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 1788
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.