Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3A Mouse anti-Human, Unconjugated, Clone: PCRP-DNMT3A-1E2, Abnova™

Catalog No. 89337408 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-408 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-408 Supplier Abnova Corporation Supplier No. MAB20913
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

DNMT3A Monoclonal antibody specifically detects DNMT3A in Human samples. It is validated for WB, IP, IHC-P.

Specifications

Antigen DNMT3A
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-DNMT3A-1E2
Conjugate Unconjugated
Description DNA (cytosine-5-)-methyltransferase 3 alpha
Gene DNMT3A
Gene Alias DNMT3A2, M.HsaIIIA
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human DNMT3A
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1788
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.