Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dnmt3L Antibody (2D10H11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261756
Description
Dnmt3L Monoclonal specifically detects Dnmt3L in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| Dnmt3L | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| DNA (cytosine-5-)-methyltransferase 3-like, DNA (cytosine-5)-methyltransferase 3-like, human cytosine-5-methyltransferase 3-like protein, 10cytosine-5-methyltransferase 3-like protein, MGC1090 | |
| Mouse | |
| 43.6 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 2D10H11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| DNMT3L | |
| Purified recombinant fragment of human DNMT3L (AA: 147-386) expressed in E. Coli. | |
| Protein G purified | |
| Chromatin Research | |
| 29947 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction