Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dnmt3L Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen Dnmt3L
Concentration 0.5 mg/mL
Dilution Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227099SS
SDP
View Documents
Novus Biologicals
NBP227099SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227099
SDP
View Documents
Novus Biologicals
NBP227099
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Dnmt3L Polyclonal specifically detects Dnmt3L in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Dnmt3L
Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml
Polyclonal
Purified
RUO
Human, Primate
Q9UJW3
29947
A portion of amino acids 25-75 from human Dnmt3L was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Chromatin Research
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
DNA (cytosine-5-)-methyltransferase 3-like, DNA (cytosine-5)-methyltransferase 3-like, human cytosine-5-methyltransferase 3-like protein, 10cytosine-5-methyltransferase 3-like protein, MGC1090
DNMT3L
IgG
Protein A purified
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, chimpanzee, and white-cheeked gibbon, 95% in macaque, and rhesus monkey, 89% in panda, cow, new world monkey, and horse, 84% in pig, and opossum, 79% in naked mole rat, elephant, rat, and mouse, and 74% homologous in Chinese hamster.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.