Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNMT3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589316
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589316 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589316 Supplier Invitrogen™ Supplier No. PA589316
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGLLQRRRKW RSQLKAFYDR ESENPLEMFE TVPVWRRQPV RVLSLFEDIK KELTSLGFLE SGSDPGQLKH VVDVTDTVRK DVEEWGPFDL VYGATPPLGH TCDRPPSWYL FQFHRLLQYA RPKPGSPRPF FWMFVDNLVL NKEDLDVASR FLEMEPVTIP DVHGGSLQNA VRVWSNIPAI RSRHWALVSE EELSLLAQNK QSSKLAAKWP TKLVKNCFLP LREYFKYFST ELTSSL; Positive Samples: HeLa, Jurkat, 293T, Mouse testis, Mouse thymus, Mouse ovary, Rat testis; Cellular Location: Nucleus.

Methylation of DNA at cytosine residues in mammalian cells is a heritable, epigenetic modification that is critical for proper regulation of gene expression, genomic imprinting, and development. Three families of mammalian DNA methyltransferases have been identified: DNMT1, DNMT2, and DNMT3. DNMT1 is constitutively expressed in proliferating cells and functions as a maintenance methyltransferase, transferring proper methylation patterns to newly synthesized DNA during replication. DNMT3A and DNMT3B are strongly expressed in embryonic stem cells with reduced expression in adult somatic tissues. DNMT3A and DNMT3B function as de novo methyltransferases that methylate previously unmethylated regions of DNA. DNMT2 is expressed at low levels in adult somatic tissues and its inactivation affects neither de novo nor maintenance DNA methylation. DNMT3L is a catalytically inactive regulatory factor for the DNMT3A and DNMT3B de novo methyltransferases that is expressed at low levels in embryonic stem cells, testis, ovaries, and thymus. These de novo methyltransferases consist of a heterotetrameric complex containing two molecules of DNMT3L, and either two molecules of DNMT3A or DNMT3B. DNMT3L contains an amino-terminal ATRX-DNMT3-DNMT3L (ADD) domain and a carboxy-terminal methyltransferase-like domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNMT3L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.06 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DNMT3L
Gene Accession No. Q9CWR8, Q9UJW3
Gene Alias cytosine-5-methyltransferase 3-like protein; D6Ertd14e; DNA (cytosine-5-)-methyltransferase 3-like; DNA (cytosine-5)-methyltransferase 3-like; DNA methyltransferase 3 like; Dnmt 3L; DNMT3L; ecat7; human cytosine-5-methyltransferase 3-like protein; MGC1090
Gene Symbols DNMT3L
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 151-386 of human DNMT3L (NP_7870631).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29947, 54427
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.