Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNMT3L Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABE291025UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE2910-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2910-25UL Supplier MilliporeSigma Supplier No. ABE291025UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-DNMT3L is a highly specific rabbit polyclonal antibody, that targets DNA (cytosine-5)-methyltransferase 3-like and has been tested in Immunofluorescence, Western Blotting and Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNMT3L
Applications Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Dnmt3l
Host Species Rabbit
Immunogen His-tagged full length (421 aa) murine recombinant DNA (cytosine-5)-methyltransferase 3-like (DNMT3L).
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Dnmt3l in murine cells.
Target Species Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.