Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNPEP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560848 Supplier Invitrogen™ Supplier No. PA560848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDRDLTLAGR VIVKCPTSGR LEQQLVHVER PILRIPHLAI HLQRNINENF GPNTEMHLVP ILATAIQEEL EKGTPEPGPL NAVDER Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

Aminopeptidase with specificity towards an acidic amino acid at the N-terminus. Likely to play an important role in intracellular protein and peptide metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNPEP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNPEP
Gene Accession No. Q9ULA0, Q9Z2W0
Gene Alias AA407814; Aspartyl aminopeptidase; ASPEP; DAP; Dnpep
Gene Symbols DNPEP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNPEP. Recombinant protein control fragment (Product #RP-95834).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13437, 23549, 301529
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.