Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNPEP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557638
Description
Immunogen sequence: SCFCALQALI DSCAGPGSLA TEPHVRMVTL YDNEEVGSES AQGAQSLLTE LVLRRISASC QHPTAFEEAI PKSFMISADM AHAVHPNYLD Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.
Aminopeptidase with specificity towards an acidic amino acid at the N-terminus. Likely to play an important role in intracellular protein and peptide metabolism.
Specifications
DNPEP | |
Polyclonal | |
Unconjugated | |
DNPEP | |
AA407814; Aspartyl aminopeptidase; ASPEP; DAP; Dnpep | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13437, 23549, 301529 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULA0, Q9Z2W0 | |
DNPEP | |
Recombinant protein corresponding to Human DNPEP. Recombinant protein control fragment (Product #RP-95833). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction