Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNTTIP1 Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
SDP

Catalog No. 502542467
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2467 30 μL
50-167-2337 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2467 Supplier OriGene Supplier No. TA501194S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNTTIP1 Monoclonal antibody specifically detects DNTTIP1 in Human samples. It is validated for Western Blot

Specifications

Antigen DNTTIP1
Applications Western Blot
Classification Monoclonal
Clone OTI1H8
Concentration 0.99 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene IGBP1
Gene Accession No. P78318
Gene Alias 6430706C13Rik; C20orf167; deoxynucleotidyltransferase; deoxynucleotidyltransferase terminal interacting protein 1; deoxynucleotidyltransferase terminal-interacting protein 1; deoxynucleotidyltransferase, terminal, interacting protein 1; dJ447F3.4; Dnttip1; RP3-447F3.5; Tdif1; TdT binding protein; tdT-interacting factor 1; terminal deoxynucleotidyltransferase interacting factor 1; terminal deoxynucleotidyltransferase-interacting factor 1
Host Species Mouse
Immunogen Full length human recombinant protein of human IGBP1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3476
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.