Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DO-11.10 Mouse, anti-Mouse, R718, Clone: KJ1-26, BD OptiBuild™

Catalog No. BDB751889 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751889 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751889 Supplier BD Biosciences Supplier No. 751889
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The KJ1-26 monoclonal antibody specifically reacts with the DO-11.10 Clonotypic T-cell Receptor (TCR) of the BALB/c-derived DO-11.10 T-cell hybridoma and T lymphocytes from the DO-11.10 transgenic mouse (TgN[DO-11.10]10Loh). The DO-11.10 TCR, an 80-90-kDa (non-reduced) or 40-44-kDa (reduced) protein, is specific for the chicken OVA(323-339)/I-A[d] complex. The DO-11.10 T-cell hybridoma also responds strongly to chicken OVA/I-A[b] and jungle fowl OVA/I-A[d] and weakly to turkey OVA/I-A[d] and I-A[b]. The DO-11.10 mouse model is valuable for studies of T-cell immigration, immunoregulation, development, activation, and function. The KJ1-26 mAb was shown to block the antigen responses of the DO-11.10 T-cell hybridoma in vitro.

TRUSTED_SUSTAINABILITY

Specifications

Antigen DO-11.10
Applications Flow Cytometry
Classification Monoclonal
Clone KJ1-26
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.