Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOC2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566787 Supplier Invitrogen™ Supplier No. PA566787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAALLGATTPE DGAEVDSYDS DDATALG Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.

There are at least two protein isoforms of the Double C2 protein, namely alpha (DOC2A) and beta (DOC2B), which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2A is mainly expressed in brain and is suggested to be involved in Ca(2+)-dependent neurotransmitter release.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOC2A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DOC2A
Gene Accession No. Q14183
Gene Alias Doc2; DOC2A; Doc2-alpha; double C2 domain alpha; double C2, alpha; double C2-like domain-containing protein alpha; double C2-like domains, alpha
Gene Symbols DOC2A
Host Species Rabbit
Immunogen Recombinant Human DOC2A. Recombinant protein control fragment (Product #RP-107445).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8448
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.