Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DOC2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100484
Description
Antibody detects endogenous levels of total DOC2B.
There are at least two protein isoforms of the Double C2 protein, namely alpha and beta, which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2B is expressed ubiquitously and is suggested to be involved in Ca-dependent intracellular vesicle trafficking in various types of cells.
Specifications
DOC2B | |
Polyclonal | |
Unconjugated | |
DOC2B | |
doc2b; doc2-beta; DOC2BL; double C2 domain beta; double C2 protein beta; double C2, beta; double C2-like domain-containing protein beta; double C2-like domain-containing protein beta; LOW QUALITY PROTEIN: double C2-like domain-containing protein beta; double C2-like domains, beta; OTTMUSP00000006522 | |
Rabbit | |
Affinity chromatography | |
RUO | |
13447, 81820, 8447 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P70169, P70610, Q14184 | |
DOC2B | |
A synthesized peptide derived from human DOC2B(Accession Q14184), corresponding to amino acid residues Q85-L135. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction