Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Docetaxel
GREENER_CHOICESDP

Catalog No. 405650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-565-0 50 mg
40-561-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-565-0 Supplier R&D Systems™ Supplier No. 4056/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Microtubule stabilizer

Binds and stabilizes microtubules; leads to cell cycle arrest and apoptosis. Displays significant antitumor activity in ovarian and gastric tumors. Prevents microtubule depolymerization and disassembly in the absence of GTP. Exhibits cytotoxic activity.

Chemical Identifiers

CAS 114977-28-5
Molecular Formula C43H53NO14

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 807.88
Purity >99%
Molecular Weight (g/mol) 807.89
Chemical Name or Material Docetaxel
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.