Learn More
Docetaxel, 99%
Description
Docetaxel is a second-generation cytotoxic antimicrotubule agent and is mostly used in the pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Docetaxel is an analog of taxol, and is considered to be a cytotoxic, semisynthetic, second-generation taxane that exhibits antineoplastic, antibiotic, and antimicrotubule activities
• Docetaxel is capable of inhibiting cellular mitosis. This compound promotes the assembly of microtubules from tubulin dimers and stabilizes microtubules by preventing depolymerization. This stability inhibits the normal dynamic reorganization of the microtubule network that is essential for vital interphase and mitotic cellular functions. This results in the persistence of abnormal microtubule structures leading to cell-cycle arrest and cell death
Application
• Docetaxel can inhibit angiogenesis by acting on vascular endothelial growth factor (VEGF) associated with cancer cells
• This compound has been used to investigate the effect of lysosomal autophagic flux in gastric cancer cells
Specifications
Specifications
| Melting Point | 186°C to 192°C |
| Color | White |
| Quantity | 100 mg |
| Merck Index | 14,3397 |
| Solubility Information | Soluble in dimethyl sulfoxide and ethanol; Insoluble in water. |
| Formula Weight | 807.88 |
| Percent Purity | 99% |
| Physical Form | Powder |
| Chemical Name or Material | Docetaxel |
Safety and Handling
| Hazard Category | H315-H319-H335-H361fd |
| Precautionary Statement | P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c |
| RTECSNumber | DA4172750 |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.