Learn More
Invitrogen™ DOCK1 Polyclonal Antibody

Description
Recommended positive controls: 293T, HeLaS3, PC-12, C8D30, NIH-3T3, Raw264.7, C2C12. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | DOCK1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.99 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | DOCK1 |
| Gene Accession No. | Q14185, Q8BUR4 |
| Gene Alias | 180 kDa protein downstream of CRK; 9130006G06Rik; AI854900; ced5; D630004B07Rik; dedicator of cytokinesis 1; dedicator of cyto-kinesis 1; dedicator of cytokinesis protein 1; dock 180; DOCK1; DOCK180; DOwnstream of CrK; RGD1566072; RP11-82L9.1 |
| Gene Symbols | DOCK1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.