Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK10 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060670JF646
Description
DOCK10 Polyclonal antibody specifically detects DOCK10 in Human samples. It is validated for Western BlotSpecifications
DOCK10 | |
Polyclonal | |
Western Blot | |
dedicator of cytokinesis 10, dedicator of cytokinesis protein 10, DKFZp781A1532, DRIP2, KIAA0694protein zizimin 3, Nbla10300, ZIZ3dopamine receptor interacting protein 2, zizimin3, zizimin-3 | |
The immunogen recognized by this antibody maps to a region between residue 2136 and 2186 of human dedicator of cytokinesis 10 using the numbering given in entry NP_055504.2 (GeneID 55619). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55619 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction