Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $728.30
Specifications
| Antigen | DOCK10 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
DOCK10 Polyclonal specifically detects DOCK10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| DOCK10 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 55619 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LDSLDNSVTCECTPEETDSSENNLHADFAKYLTETEDTVKTTRNMERLNLFSLDPDIDTLKLQKKDLLEPESVIKPFEEKAAKRIMIICKALNSN | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| dedicator of cytokinesis 10, dedicator of cytokinesis protein 10, DKFZp781A1532, DRIP2, KIAA0694protein zizimin 3, Nbla10300, ZIZ3dopamine receptor interacting protein 2, zizimin3, zizimin-3 | |
| DOCK10 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title