Learn More
DOCK10 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301306AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Potential guanine nucleotide exchange factor (GEF). GEF proteins activate some small GTPases by exchanging bound GDP for free GTP.Specifications
| DOCK10 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q96BY6 | |
| DOCK10 | |
| Between 2136 and 2186 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| DOCK10 | |
| dopamine receptor interacting protein 2, DRIP2, KIAA0694, Nbla10300, ZIZ3, zizimin3, Zizimin-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55619 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.