Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK11 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10081662AF700
Description
DOCK11 Polyclonal antibody specifically detects DOCK11 in Human samples. It is validated for Western BlotSpecifications
DOCK11 | |
Polyclonal | |
Western Blot | |
ACG, Activated Cdc42-associated guanine nucleotide exchange factor, Activated Cdc42-associated guanine nucleotide exchange factor, ACG, bB128O4.1, bB128O4.1.1 (novel protein), dedicator of cytokinesis 11, FLJ32122, FLJ43653, ZIZ2dedicator of cytokinesis protein 11, zizimin2, zizimin-2 | |
The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human dedicator of cytokinesis 11 using the numbering given in entry NP_653259.3 (GeneID 139818). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
139818 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction