Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | DOCK11 |
---|---|
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
DOCK11 Polyclonal specifically detects DOCK11 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
DOCK11 | |
Polyclonal | |
Rabbit | |
Human | |
ACG, Activated Cdc42-associated guanine nucleotide exchange factor, Activated Cdc42-associated guanine nucleotide exchange factor, ACG, bB128O4.1, bB128O4.1.1 (novel protein), dedicator of cytokinesis 11, FLJ32122, FLJ43653, ZIZ2dedicator of cytokinesis protein 11, zizimin2, zizimin-2 | |
DOCK11 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
139818 | |
This antibody was developed against a recombinant protein corresponding to amino acids: KDTVETAQDDETSSQGKAENIMASLERSMHPELMKYGRETEQLNKLSRGDGRQNLFSFDSEVQRLDFSGIEPDIKPFEEKCNKRFLVNCHDLTF | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title