Learn More
DOCK180 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301288A

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene product binds to the SH3 domain of CRK protein. It may regulate cell surface extension and may have a role in the cell surface extension of an engulfing cell around a dying cell during apoptosis.Specifications
DOCK180 | |
Polyclonal | |
Unconjugated | |
DOCK1 | |
RP11-82L9.1, DOCK180, ced5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1793, 330662 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q14185, Q8BUR4 | |
Dock1 | |
Between 1815 and 1865 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.