Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK2 Antibody (OTI7G2), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270583APC
Description
DOCK2 Monoclonal antibody specifically detects DOCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
DOCK2 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
1794 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI7G2 | |
Western Blot, Immunohistochemistry | |
dedicator of cytokinesis 2, dedicator of cytokinesis protein 2, FLJ46592, KIAA0209dedicator of cyto-kinesis 2 | |
Human recombinant protein fragment corresponding to amino acids 1544-1830 of human DOCK2 (NP_004937) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction