Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK2 Antibody (OTI7G2), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246468
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
DOCK2 Monoclonal specifically detects DOCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DOCK2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150 | |
dedicator of cytokinesis 2, dedicator of cytokinesis protein 2, FLJ46592, KIAA0209dedicator of cyto-kinesis 2 | |
Mouse | |
Immunogen affinity purified | |
RUO | |
1794 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Immunohistochemistry (Paraffin), Immunohistochemistry | |
OTI7G2 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
DOCK2 | |
Human recombinant protein fragment corresponding to amino acids 1544-1830 of human DOCK2 (NP_004937) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction