Learn More
Invitrogen™ DOCK2 Monoclonal Antibody (OTI7G2)

Description
DOCK2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | DOCK2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI7G2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DOCK2 |
| Gene Accession No. | Q92608 |
| Gene Alias | AI662014; AW122239; CED-5; dedicator of cytokinesis 2; dedicator of cyto-kinesis 2; dedicator of cytokinesis protein 2; DOCK2; FLJ46592; Hch; IMD40; KIAA0209; LOW QUALITY PROTEIN: dedicator of cytokinesis protein 2; MBC; Protein Hch |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.