Learn More
DOCK2 Mouse anti-Human, Clone: OTI7G2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802698

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The DOCK2 gene encodes a hematopoietic cell-specific CDM family protein that is indispensable for lymphocyte chemotaxis.Specifications
DOCK2 | |
Monoclonal | |
Unconjugated | |
DOCK2 | |
IMD40 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1794 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7G2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q92608 | |
DOCK2 | |
Human recombit protein fragment corresponding to amino acids 1544-1830 of human DOCK2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.