Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOCK2 Mouse anti-Human, Clone: UMAB142, liquid, UltraMAB™
SDP

Catalog No. 501685491
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-5491 30 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-5491 Supplier OriGene Supplier No. UM870039
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The DOCK2 gene encodes a hematopoietic cell-specific CDM family protein that is indispensable for lymphocyte chemotaxis.

Specifications

Antigen DOCK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone UMAB142
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DOCK2
Gene Accession No. Q92608
Gene Alias IMD40
Gene Symbols DOCK2
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1544-1830 of human DOCK2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1794
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.