Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DOCK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100485
Description
Antibody detects endogenous levels of total DOCK3;MOCA.
Dedicator of cytokinesis3 (DOCK3), also named as MOCA and PBP, is a ~180 kDa protein involved in signaling transduction. It is a potential guanine nucleotide exchange factor (GEF) which activate some small GTPases by exchanging bound GDP for free GTP. DOCK3 is associated in Alzheimer disease tangles and regulates the accumulation of amyloid precursor protein and beta-amyloid. Overexpression of Dock3 in neural cells promotes axonal outgrowth downstream of brain-derived neurotrophic factor (BDNF) signaling. DOCK3 binds to and inactivates glycogen synthase kinase-3beta (GSK-3beta) at the plasma membrane, thereby promoting axon branching and microtubule assembly. By stimulating actin polymerization and microtubule assembly, DOCK3 plays important roles downstream of BDNF signaling in the CNS.
Specifications
DOCK3 | |
Polyclonal | |
Unconjugated | |
Dock3 | |
dedicator of cytokinesis 3; dedicator of cyto-kinesis 3; dedicator of cytokinesis protein 3; DOCK3; dok-3; KIAA0299; mKIAA0299; MOCA; Modifier of cell adhesion; PBP; Presenilin-binding protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
1795, 208869 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8CIQ7, Q8IZD9 | |
Dock3 | |
A synthesized peptide derived from human DOCK3(Accession Q8IZD9), corresponding to amino acid residues C550-S600. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction