Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK4 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP126649AF647
Description
DOCK4 Polyclonal antibody specifically detects DOCK4 in Human samples. It is validated for Western BlotSpecifications
DOCK4 | |
Polyclonal | |
Western Blot | |
dedicator of cytokinesis 4, dedicator of cytokinesis protein 4, KIAA0716FLJ34238, MGC134911, MGC134912 | |
The immunogen recognized by this antibody maps to a region between residue 1916 and 1966 of human dedicator of cytokinesis 4 using the numbering given in entry NP_055520.3 (GeneID 9732). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9732 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction