Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOCK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567177
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567177 Supplier Invitrogen™ Supplier No. PA567177
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSAIYPTPVEP SQRMLFNHIG DGALPRSDPN LSAPEKAVNP TPSSWSLDSG KEAKNMSDSG KLISP Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

The DOCK4 gene encodes the dedicator of cytokinesis 4 protein, a member of the DOCK family, which functions as a guanine nucleotide exchange factor (GEF) for small Rho GTPases, specifically Rac1 and Rap1. DOCK4 is critical for regulating various cellular processes, including cell migration, polarity, and cytoskeletal organization. It plays a significant role in maintaining the integrity of endothelial barriers and mediating intercellular junctions. Mutations or altered expression of DOCK4 have been implicated in cancer progression, particularly in tumor metastasis and invasion, by affecting the adhesion and migration of cancer cells. Additionally, DOCK4 is involved in neurodevelopmental processes and has been associated with psychiatric disorders, reflecting its importance in brain function and development. Understanding the roles of DOCK4 in both health and disease could provide insights into therapeutic strategies for cancer and neurological disorders where these pathways are disrupted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOCK4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DOCK4
Gene Accession No. Q8N1I0
Gene Alias 5330406C03; 6330411N01Rik; AF263288; C030023J22; dedicator of cytokinesis 4; dedicator of cytokinesis protein 4; DOCK4; Kiaa0716; mKIAA0716; RGD1561724
Gene Symbols DOCK4
Host Species Rabbit
Immunogen Recombinant Human DOCK4. Recombinant protein control fragment (Product #RP-107821).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9732
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.