Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ DOCK4 Polyclonal Antibody
DOCK4 Polyclonal Antibody
Supplier: Thermo Scientific™ PA5109224
Description
DOCK4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
DOCK4 | |
Polyclonal | |
Unconjugated | |
DOCK4 | |
5330406C03; 6330411N01Rik; AF263288; C030023J22; dedicator of cytokinesis 4; dedicator of cytokinesis protein 4; DOCK4; Kiaa0716; mKIAA0716; RGD1561724 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9732 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N1I0 | |
DOCK4 | |
Synthetic peptide within Human DOCK4 aa 1,917-1,966. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction