Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOCK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566045
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566045 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566045 Supplier Invitrogen™ Supplier No. PA566045
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VELSVLFCKFI QSIPDNQLVR QKLNCMTKIV ESTLFRQSEC REVLLPLLTD QLSGQLDDNS NKPDHEASSQ LL Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.

Guanine nucleotide exchange factor (GEF) for Rho and Rac. GEF proteins activate small GTPases by exchanging bound GDP for free GTP. Along with DOCK1, mediates CRK/CRKL regulation of epithelial and endothelial cell spreading and migration on type IV collagen (PubMed:19004829).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOCK5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DOCK5
Gene Accession No. Q9H7D0
Gene Alias dedicator of cytokinesis 5; dedicator of cytokinesis protein 5; DOCK5; FLJ21034
Gene Symbols DOCK5
Host Species Rabbit
Immunogen Recombinant Human DOCK5. Recombinant protein control fragment (Product #RP-106733).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80005
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.