Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK7 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178748AF532
Description
DOCK7 Polyclonal antibody specifically detects DOCK7 in Human samples. It is validated for Western BlotSpecifications
DOCK7 | |
Polyclonal | |
Western Blot | |
dedicator of cytokinesis 7, dedicator of cytokinesis protein 7, KIAA1771ZIR2 | |
The immunogen for this product maps to a region between residue 175 and 225 of human Dedicator of Cytokinesis 7 using the numbering given in entry Q96N67.4 (GeneID 85440). | |
0.1 mL | |
Cell Biology, GPCR, Neuroscience, Signal Transduction | |
85440 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction