Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK8 Monoclonal Antibody (OTI5C4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA506495

Description
The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors. DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse. Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome.Specifications
DOCK8 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8NF50 | |
DOCK8 | |
Human recombit protein fragment corresponding to amino acids 833-1160 of human DOCK8 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI5C4 | |
Unconjugated | |
DOCK8 | |
HEL-205, MRD2, ZIR8 | |
Mouse | |
Affinity Chromatography | |
RUO | |
81704 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction