Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOCK8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578589 Supplier Invitrogen™ Supplier No. PA578589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Raji, NCI-H929 Predicted Reactivity: Rat (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors. DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse. Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOCK8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Dock8
Gene Accession No. Q8C147, Q8NF50
Gene Alias 1200017A24Rik; 5830472H07Rik; A130095G14Rik; AI461977; dedicator of cytokinesis 8; dedicator of cytokinesis protein 8; Dock8; epididymis luminal protein 205; HEL-205; MRD2; ZIR8
Gene Symbols Dock8
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human DOCK8. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76088, 81704
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.