Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOCK8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140262 Supplier Invitrogen™ Supplier No. PA5140262
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: INRYSSAEIR KQFTLPPNLG QYHRQSISTS GFPSLQLPQF YDPVEPVDFE GLLMTHLNSL DVQLAQELGD FTDDDLDVVF Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 86%.

The Dedicator of cytokinesis protein 8 (DOCK8) is a member of the DOCK180 family of guanine nucleotide exchange factors. DOCK8 plays an essential role in humoral immune responses and is important in the proper formation of the B cell immunological synapse. Mutations in this gene result in the autosomal recessive form of the hyper-IgE syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOCK8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dock8
Gene Accession No. Q8NF50
Gene Alias 1200017A24Rik; 5830472H07Rik; A130095G14Rik; AI461977; dedicator of cytokinesis 8; dedicator of cytokinesis protein 8; Dock8; epididymis luminal protein 205; HEL-205; MRD2; ZIR8
Gene Symbols Dock8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DOCK8. Recombinant protein control fragment (Product #RP-109342).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81704
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.