Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DOCK9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | DOCK9 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2307
![]() ![]() |
Proteintech Group Inc
189871AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Formin-like protein 2 is a cytoplasmic phosphoprotein belonging to the formin homology domain containing protein family and has a DAD (diaphanous autoregulatory domain), FH2 (formin homology 2) domain, and GBD/FH3 (Rho GTPase-binding/formin homology 3) domain. The exact function of FMNL2 is yet to be known but reports suggest that it may have a role in the Wnt signaling pathway. Formin-related proteins are also known to play an important role in morphogenesis, cytokinesis and cell polarity. Several alternatively spliced transcript variants have been described but their full-length nature is not yet determined. It is also known to be associated with a diffuse-type gastric cancer, breast cancer, chondrosarcoma, melanoma, and glioblastoma. FMNL2 might be implicated in polarity control, invasion, migration or metastasis through actin filament nucleation and may be regulated by Rho family GTPase.Specifications
DOCK9 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8BIK4, Q9BZ29 | |
105445, 23348 | |
Peptide | |
Primary | |
-20°C | |
DOCK9 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
dedicator of cytokinesis 9, DOCK9, KIAA1058, KIAA1085, ZIZ1, ZIZIMIN1 | |
Dock9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title