Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doctor Solution (Sodium Plumbite) ASTM D 4952, Reagents

Doctor Solution (Sodium Plumbite) ASTM D 4952, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

$80.90 - $234.50

Specifications

Chemical Name or Material Doctor Solution (Sodium Plumbite) ASTM D 4952
Color Colorless with Slight Orange
CAS 7732-18-5, 1310-73-2
Grade Reagent
Packaging Poly Bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
01-335-432
Accessories View Documents
Reagents Holdings Llc
CD1001001A
1 L
Each for $80.90
Only null left
Add to Cart
 
01-335-431
Accessories View Documents
Reagents Holdings Llc
CD1001004A
4 L
Each for $234.50
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Doctor Solution (Sodium Plumbite) ASTM D 4952
7732-18-5, 1310-73-2
Poly Bottle
Non-sterile
Colorless with Slight Orange
Reagent
1.14
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.