Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Traceable™ Digital Thermometer with Stainless-Steel Stem, 0.25 in. LCD Screen, and Protective Guard
Shop All Fisherbrand Products
Click to view available options
Temperature Range (Metric):
-50°C to +150°C
-50°C to +250°C
-50°C to +280°C
-50°C to +300°C
Temperature Range (English):
-58°F to +302°F
-58°F to +482°F
-58°F to +536°F
-58°F to +572°F
Accuracy:
±0.2°C
±0.4°C
±0.5°C
±1°C
±1°C (-20°C to +120°C)
±1.5°C
Length (Metric) Probe:
8.9 cm
20 cm
20.32 cm
28.87 cm
Description
Easy-to-read handheld Traceable™ Digital Thermometer.
- Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
- Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
- For use in cuvets, soil, incubators, petroleum products and foods
Certifications
Multi-point calibration on individually-numbered Traceable Certificate assures accuracy from ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA; indicates traceability to NIST.
Specifications
Specifications
Certifications/Compliance | DNV, ISO 17025, A2LA |
Diameter (English) Probe | 0.14 in. |
Diameter (Metric) Probe | 3.56 mm |
For Use With (Application) | Read temperatures in foods, gases, samples, reagents, water baths and semisolids |
Includes | Traceable certificate, Stainless steel probe |
Length (English) Probe | 11.37 in. |
Length (Metric) | 365 mm |
Length (Metric) Probe | 28.87 cm |
Material | ABS Plastic |
Resolution | 0.1°C |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction