Learn More
Dodecacarbonyltriiron, 96% (dry wt.), stab. with 5-10% methanol
Description
It eliminates sulfur from substituted thiophene complexes, which leads to ferroles. Access to new areas of organometallic chemistry is promising using the reaction of Fe3(CO)12 with tellurium-nitrogen heterocycles such as benzoisotellurazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 140°C (decomposition) |
Color | Black |
Density | 2.00 g/mL |
Linear Formula | Fe3(CO)12 |
Quantity | 10 g |
UN Number | UN3179 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. Soluble in non polar organic solvents. |
Formula Weight | 503.67 |
Percent Purity | 96% |
Show More |
Safety and Handling
Hazard Category | H228-H302+H312+H332-H371 |
Hazard Statement | GHS H Statement H228-H331-H371-H302-H312 Flammable solid. Toxic if inhaled. May cause damage to organs. Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P210-P240-P241-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P309+P311-P312-P330-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S. |
EINECSNumber | 241-668-5 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.