Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecacarbonyltriiron, 96% (dry wt.), stab. with 5-10% methanol

Catalog No. p-7049152
Click to view available options
Quantity:
2 g
10 g
50 g

Reducing agent (reduces NO2), catalyst for alkyne trimerization | CAS: 17685-52-8

It eliminates sulfur from substituted thiophene complexes, which leads to ferroles. Access to new areas of organometallic chemistry is promising using the reaction of Fe3(CO)12 with tellurium-nitrogen heterocycles such as benzoisotellurazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17685-52-8
MDL Number MFCD00135617
Synonym triiron dodecarbonyl

Specifications

Melting Point 140°C (decomposition)
Color Black
Density 2.00 g/mL
Linear Formula Fe3(CO)12
Quantity 2 g
UN Number UN3179
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water. Soluble in non polar organic solvents.
Formula Weight 503.67
Percent Purity 96%
Physical Form Crystalline
Assay (dry wt.)
Chemical Name or Material Dodecacarbonyltriiron, Stabilized with 5 to 10% methanol
Show More Show Less
Hazard Category H228-H302+H312+H332-H371
Hazard Statement GHS H Statement
H228-H331-H371-H302-H312
Flammable solid.
Toxic if inhaled.
May cause damage to organs.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P240-P241-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P309+P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
EINECSNumber 241-668-5
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.