Alfa Aesar™ Dodecacarbonyltriiron, 96% (dry wt.), stab. with 5-10% methanol

Reducing agent (reduces NO2), catalyst for alkyne trimerization

Manufacturer: Alfa Aesar™ 1305904

Quantity 2g

 View more versions of this product

Catalog No. AA1305904

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17685-52-8
    Linear Formula Fe3(CO)12
    Formula Weight 503.67
    MDL Number MFCD00135617
    UN Number UN3179
    Synonym Triiron dodecacarbonyl; Iron dodecacarbonyl
    Chemical Name or Material Dodecacarbonyltriiron, Stabilized with 5 to 10% methanol

    Specifications

    Formula Weight 503.67
    Health Hazard 3 GHS P Statement
    P210-P280h-P309-P310-P404
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a closed container.
    MDL Number MFCD00135617
    UN Number UN3179
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H228-H331-H371-H302-H312
    Flammable solid.
    Toxic if inhaled.
    May cause damage to organs.
    Harmful if swallowed.
    Harmful in contact with skin.
    Density 2.00g/mL
    Synonym Triiron dodecacarbonyl; Iron dodecacarbonyl
    Melting Point 140°C (dec.)
    Sensitivity Air & Moisture Sensitive
    Physical Form Crystalline
    Linear Formula Fe3(CO)12
    CAS 17685-52-8
    Color Black
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.
    Quantity 2g
    Chemical Name or Material Dodecacarbonyltriiron, Stabilized with 5 to 10% methanol
    Assay 96%, (dry wt.)

    GHS H Statement
    H228-H331-H371-H302-H312
    Flammable solid.
    Toxic if inhaled.
    May cause damage to organs.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P210-P280h-P309-P310-P404
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit