Alfa Aesar™ Dodecacarbonyltriosmium, 99%

Manufacturer: Alfa Aesar™ 1303804

Quantity 2g

 View more versions of this product

Catalog No. AA1303804

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15696-40-9
    Formula Weight 906.73
    MDL Number MFCD00011149
    UN Number UN3466
    Synonym Triosmium dodecacarbonyl; Osmium carbonyl
    Chemical Name or Material Dodecacarbonyltriosmium

    Specifications

    Formula Weight 906.73
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Crystalline
    CAS 15696-40-9
    UN Number UN3466
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H330
    Fatal if swallowed.
    Fatal if inhaled.
    MDL Number MFCD00011149
    Synonym Triosmium dodecacarbonyl; Osmium carbonyl
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
    Quantity 2g
    Melting Point 224°C
    Chemical Name or Material Dodecacarbonyltriosmium
    Assay 99%

    GHS H Statement
    H300-H330
    Fatal if swallowed.
    Fatal if inhaled.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit