Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecanal, 95%, stab.

Catalog No. p-7022716
Click to view available options
Quantity:
100 g
500 g

Store cold | CAS: 112-54-9 | C12H24O | 184.32 g/mol

Dodecyl aldehyde is used as a fragrance agent. It is used as a common ingredient in perfumery. It's end applications include soap, detergent, beauty care product, household product.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecyl aldehyde is used as a fragrance agent. It is used as a common ingredient in perfumery. It′s end applications include soap, detergent, beauty care product, household product.

Solubility
Soluble in chloroform, methanol, alcohol, and propylene glycol. Insoluble in glycerol and water.

Notes
Air sensitive. Store under dry inert gas. Store in cool dry place in tightly closed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-54-9
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.32
MDL Number MFCD00007017
InChI Key HFJRKMMYBMWEAD-UHFFFAOYSA-N
Synonym lauraldehyde, dodecyl aldehyde, lauric aldehyde, lauryl aldehyde, n-dodecanal, dodecanaldehyde, 1-dodecanal, n-dodecyl aldehyde, n-lauraldehyde, n-dodecylic aldehyde
PubChem CID 8194
ChEBI CHEBI:27836
IUPAC Name dodecanal
SMILES CCCCCCCCCCCC=O

Specifications

Melting Point 12°C
Density 0.835
Boiling Point 184°C to 186°C (100 mm)
Flash Point 101°C (214°F)
Assay Percent Range 95%
Refractive Index 1.435
Quantity 100 g
Beilstein 1703917
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,methanol,alcohol,and propylene glycol. Insoluble in glycerol and water.
Formula Weight 184.32
Percent Purity 95%
Chemical Name or Material Dodecanal, stabilized with 250ppm BHT
Show More Show Less
Hazard Category H315-H317-H319
Hazard Statement GHS H Statement
H315-H319-H335
Precautionary Statement P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-983-6
RTECSNumber JR9100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.