Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecane, 99%, pure

Catalog No. p-124400
Change view
Click to view available options
Quantity:
25 mL
100 mL
250 mL
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117590250 25 mL Glass bottle
AC117591000 100 mL Glass bottle
AC117592500 250 mL Glass bottle
AC117595000 500 mL Glass bottle
AC117590025 2.5 L Glass bottle
AC117590100 10 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. AC117590250 Supplier Thermo Scientific Chemicals Supplier No. 117590250
Only null left
Add to Cart

CAS: 112-40-3 | C12H26 | 170.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -12.0°C
Density 0.7530g/mL
Boiling Point 215.0°C to 217.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C12H26
Refractive Index 1.4210 to 1.4230
Linear Formula CH3(CH2)10CH3
Quantity 25 mL
Beilstein 01, 171
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Vapor Pressure 1mbar at 48°C
Formula Weight 170.34
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material n-Dodecane, 99%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement Emergency Overview
Combustible liquid. Harmful by inhalation. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking.
NFPA
EINECSNumber 203-967-9
RTECSNumber JR2125000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.