Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecane, ≥99%, Reagent Grade, Solstice

Catalog No. 60047947 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-947 2.5 L Glass bottle
60-047-946 100 mL Glass bottle
60-047-948 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-047-947 Supplier Solstice Supplier No. D2211042.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -9.6°C
Color Colorless
Density 0.75g/mL at 25°C
Boiling Point 215°C to 217°C
Odor Characteristic
Packaging Glass bottle
Linear Formula CH3(CH2)10CH3
Quantity 2.5 L
Beilstein 1697175
Vapor Density 5.96 (vs air)
Formula Weight 170.33g/mol
Percent Purity ≥99%
Chemical Name or Material Dodecane
Show More Show Less
Hazard Statement P301 + P310-P331
RTECSNumber 203-967-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.